Non-Coded Lumina Lamp Plug Adapter, PerkinElmer
Supplier: PerkinElmer
N3050198
97013-406EA 247.47 USD
97013-406
Non-Coded Lumina Lamp Plug Adapter, PerkinElmer
Lights Hollow Cathode Lamps
Allows Lumina™ HCLs (N305XXXX series, 4-pin) to be used with , PerkinElmer atomic absorption instruments with non-coded lamp connectors (9-pin).
PerkinElmer also offers lamp adapters that are coded to specific element(s).