Lumina™ Vanadium (V) Hollow Cathode Lamp, PerkinElmer
Supplier: PerkinElmer
N3050186
97013-390EA 1602.62 USD
97013-390
Lumina™ Vanadium (V) Hollow Cathode Lamp, PerkinElmer
Lights Hollow Cathode Lamps
Single-element Lumina™ Hollow Cathode Lamp (HCL) for the detection of elemental Vanadium (V). Series N30501XX Lumina™ 2" (50mm) diameter single-element lamps are designed to be used with PerkinElmer PinAAcle™ and AAnalyst™ Atomic Absorption (AA) spectrometer series of instruments.
  • Lumina™ Hollow Cathode Lamps may also be used with all earlier AA spectrometers that do not have automatic code reading capability by using an appropriate adapter cable
  • This element requires the use of a nitrous oxide-acetylene flame
Every genuine PerkinElmer lamp is designed for use with and tested on spectrometers to assure compatibility and high level performance. PerkinElmer offers a wide range of single-element and multi-element HCLs, which are ideal for determining most elements by atomic absorption spectroscopy. To prolong the life of a Lumina™ Hollow Cathode Lamp (HCL), they are produced with larger internal volume, providing a greater supply of fill gas at optimum pressure. The larger the lamp, the greater the inert gas volume, and the longer the lamp lifetime.