Xenon Source Lamps for PerkinElmer Fluorescence and Luminescence Spectrometers, PerkinElmer
Supplier: PerkinElmer
L2251157
97013-380EA 2033.01 USD
97013-380
Xenon Source Lamps for PerkinElmer Fluorescence and Luminescence Spectrometers, PerkinElmer
Lights Xenon Lights, Lamps and Bulbs
Sources for the PerkinElmer LC-240 fluorescence detector and luminescence spectrometers are pulsed xenon discharge lamps (7.3 W average power at 50 Hz).
This reliable source produces very little ozone or heat but provides remarkable sensitivity due to its peak intensity exceeding 1 kW. On most LS Series instruments the detector electronics are controlled so that delay and gate time can be varied to measure phosphorescence between source pulses.
Caution: Xenon lamp quartz envelopes are under pressure. Extreme caution and eye protection are necessary.