Potassium (K) - Sodium (Na) Lumina™ Hollow Cathode Lamp, PerkinElmer
Supplier: PerkinElmer
N3050204
97013-340EA 2036.21 USD
97013-340
Potassium (K) - Sodium (Na) Lumina™ Hollow Cathode Lamp, PerkinElmer
Lights Sodium Lamps
Series N30502XX Lumina™ 50 mm (2") diameter single-element lamps are designed to be used with our PinAAcle™ and AAnalyst™ Atomic Absorption spectrometer series of instruments.
Multi-Element Lumina™ Hollow Cathode Lamp (HCL) for the detection of elemental Potassium (K) and Sodium (Na). Please note that multi-element lamps are not designed to be used in a furnace system. Every genuine PerkinElmer lamp is designed for use with and tested on our spectrometers to assure compatibility and the highest performance. We offer a wide range of single-element and multi-element HCLs, which are ideal for determining most elements by atomic absorption spectroscopy. To prolong the life of a Lumina™ Hollow Cathode Lamp (HCL), we produce lamps with larger internal volume so that a greater supply of fill gas at optimum pressure is available. The larger the lamp, the greater the inert gas volume - and the longer the lamp lifetime. Note: Lumina™ Hollow Cathode Lamps may also be used with all earlier AA spectrometers that do not have automatic code reading capability by using an appropriate adapter cable.