UV/VIS Deuterium Detector Lamp for PerkinElmer LC Instruments, PerkinElmer
Supplier: PerkinElmer
N2920149
97013-314EA 1730.5 USD
97013-314
UV/VIS Deuterium Detector Lamp for PerkinElmer LC Instruments, PerkinElmer
Lights Deuterium Lamps
Our extensive quality control and inspection process demands the very best quality sources. Choosing a deuterium source provides outstanding ultraviolet and true visible performance.
  • Lamp changes are quick and easy due to a unique self-aligning lamp mount.
  • Exceptional performance anywhere in the detectors 190 to 700 nm wavelength range
  • Maintain factory performance using genuine replacement parts