Aluminum (Al) - Calcium (Ca) - Magnesium (Mg) Lumina™ Hollow Cathode Lamp, PerkinElmer
Supplier: PerkinElmer
N3050207
97013-272EA 8203.8 USD
97013-272
Aluminum (Al) - Calcium (Ca) - Magnesium (Mg) Lumina™ Hollow Cathode Lamp, PerkinElmer
Lights Hollow Cathode Lamps
Multi-Element Lumina™ Hollow cathode lamp (HCL) for the detection of elemental Aluminum (Al), Calcium (Ca), and Magnesium (Mg). Series N30502XX Lumina™ 2" (50 mm) diameter multi-element lamps are designed to be used with our PinAAcle™ and AAnalyst™ Atomic Absorption spectrometer series of instruments.
Please note that multi-element lamps are not designed to be used in a furnace system. Every genuine , PerkinElmer lamp is designed for use with and tested on our spectrometers to assure compatibility and the highest performance. We offer a wide range of single-element and multi-element HCLs, which are ideal for determining most elements by atomic absorption spectroscopy. To prolong the life of a Lumina™ Hollow Cathode Lamp (HCL), we produce lamps with larger internal volume so that a greater supply of fill gas at optimum pressure is available. The larger the lamp, the greater the inert gas volume - and the longer the lamp lifetime.
Caution: Note: Lumina™ Hollow Cathode Lamps may also be used with all earlier AA spectrometers that do not have automatic code reading capability by using an appropriate adapter cable.