Lumina™ Multi-Element Hollow Cathode Lamp, PerkinElmer
Supplier: PerkinElmer
N3050213
97013-266EA 2036.21 USD
97013-266
Lumina™ Multi-Element Hollow Cathode Lamp, PerkinElmer
Lights Hollow Cathode Lamps
Series N30502XX Lumina™ 50 mm (2") diameter multi-element lamps are designed to be used with PerkinElmer PinAAcle™ and AAnalyst™ Atomic Absorption spectrometer series of instruments.
Multi-Element Lumina™ Hollow Cathode Lamp (HCL) for the detection of elemental Silver (Ag), Chromium (Cr), Copper (Cu), Iron (Fe), and Nickel (Ni).
Lumina™ hollow cathode lamps may also be used with all earlier AA spectrometers that do not have automatic code reading capability by using an appropriate adapter cable.
Caution: Please note that multi-element lamps are not designed to be used in a furnace system.