Quick Press, PerkinElmer
Supplier: PERKINELMER U.S. LLC
01860436
97013-212EA 3881.6 USD
97013-212
Quick Press, PerkinElmer
Presses
The quick press consists of a handpress, a 7 mm die set, and a metal collar.
  • Stainless steel construction
  • Ergonomic handle design
  • Easy to install
Micropellets can be prepared in the quick press by purchasing the optional 3 mm or 1 mm die. The KBr pellet is formed in a metal collar that fits into an adjustable pellet holder.