In-line Filter Traps, PerkinElmer
Supplier: PerkinElmer
N9301178
97013-134EA 720.96 USD
97013-134
In-line Filter Traps, PerkinElmer
Filters In-Line Filters
Pre-instrument in-line filter trap for removal of moisture, oil, and dust from GC input gases.