Balston Type 95A Acetylene Filter, PerkinElmer
Supplier: PERKINELMER U.S. LLC
N9301399
97013-124EA 1028.61 USD
97013-124
Balston Type 95A Acetylene Filter, PerkinElmer
Filters Gas Filters
Filters remove particulate and acetone droplets from acetylene to 99.99% at 0.1 µm, protecting microcomputer gas controls and AA burner systems from contamination and corrosion.
Recommended for use with AA spectrometers, where the air/acetylene filter is not installed. Includes mounting bracket and appropriate fittings for installation on an acetylene line.