Thermogreen® Injector Septa, PerkinElmer
Supplier: PERKINELMER U.S. LLC
N6621028
97013-078PK 333.39 USD
97013-078
Thermogreen® Injector Septa, PerkinElmer
Septa
Thermogreen® Injector Septa are extremely low bleed over a wide range of inlet temperatures from 100 to 350 °C.
  • Functional over a wide range of temperatures
Easier needle penetration and high puncture tolerance make this septum ideal for autosamplers. Septa are already conditioned and ready to use.