Copper Torch Ignitor Tape for Optima™ 2x00/4x00/5x00/7x00 DV, PerkinElmer
Supplier: PerkinElmer
N0775297
97013-028EA 64.06 USD
97013-028
Copper Torch Ignitor Tape for Optima™ 2x00/4x00/5x00/7x00 DV, PerkinElmer
Tapes
The copper foil torch ignitor tape is used with Optima™ 2x00 DV, 4x00 DV, 5x00 DV, and 7x00 DV ICP-OES series instruments. The roll of tape includes 120 pieces.
This copper ignitor tape is also included in the scott/cross-flow II sample introduction kit for Optima™ 2x00/4x00/5x00/7x00 DV (101692-222), the cyclonic/concentric sample introduction kit for Optima™ 2x00/4x00/5x00/7x00 DV (101676-468), and the organics kit for Optima™ 4300 V/5300 V/7300 V(10136-316).