GemClean Cross-Flow II Nebulizer, PerkinElmer
Supplier: PERKINELMER U.S. LLC
N8120515
97013-026PK 412.75 USD
97013-026 89260-276
GemClean Cross-Flow II Nebulizer, PerkinElmer
Atomizers
Complete chemically inert Cross-Flow II nebulizer assembly for ICP-MS. Specially processed to reduce BEC levels below 1 ppb.
  • Suitable for NexION/ELAN DRCe/9000/6X00/5000/500/250
Contains an end cap with a GemTip clear sapphire sample tip and a GemTip red ruby argon tip optimized for ICP-MS.