Silanized Glass Wool, PerkinElmer
Supplier: PERKINELMER U.S. LLC
00097993
97012-952EA 288.94 USD
97012-952
Silanized Glass Wool, PerkinElmer
Glass and Cotton Wool
Silanized glass wool for automated thermal desorber 400.
  • Silanized for general-purpose use
  • Nonflammable
  • Convenient packing material
Glass wool is made up of short glass fibers and is nonflammable in nature. It is widely used as thermal insulation material in ships, automobiles, air conditioning ducts, and water supply pipes.