Demountable Quartz Torch, 0-Slot for Optima™ 3000 SCX/3x00 XL, PerkinElmer
Supplier: PERKINELMER U.S. LLC
N0695379
97012-916EA 1024.26 USD
97012-916
Demountable Quartz Torch, 0-Slot for Optima™ 3000 SCX/3x00 XL, PerkinElmer
Chromatography Supplies Chromatography Spare Parts
This quartz torch is designed for use with Optima™ 3000 SCX and 3x00 XL ICP-OES series instruments.
This demountable 0-slot torch design uses one-piece quartz tubing for plasma and auxiliary gas flow. It is recommended for use with organic solvents.