Torch Bonnet, PerkinElmer
Supplier: PerkinElmer
N0775289
97012-906EA 365.79 USD
97012-906
Torch Bonnet, PerkinElmer
Lights Flashlights
This quartz bonnet is used with Optima™ 2x00 DV, 4x00 DV, 5x00 DV, 7x00 DV ICP-OES series instruments.