Radial Purge Extension Window for PerkinElmer Avio and Optima ICP-OES Instruments, PerkinElmer
Supplier: PerkinElmer
N0690672
97012-902EA 326.57 USD
97012-902
Radial Purge Extension Window for PerkinElmer Avio and Optima ICP-OES Instruments, PerkinElmer
Optical Windows
This radial purge extension window, made of quartz, is used with Avio® 200 and Optima® 2x00, 7000 DV, and 8000 ICP-OES series instruments.
The radial view purge extension window is a replaceable quartz window that slides into the purge extension between the torch and the optics.