Demountable Quartz Torch, 3-Slot, for Optima ICP-OES Instruments, PerkinElmer
Supplier: PerkinElmer
N0771500
97012-866EA 722 USD
97012-866
Demountable Quartz Torch, 3-Slot, for Optima ICP-OES Instruments, PerkinElmer
Lights Flashlights
This 3-Slot quartz torch is used with Optima™ 4300, 5300, and 7300 V ICP-OES series instruments.
This demountable 3-slot torch design uses one-piece quartz tubing for plasma and auxiliary gas flow and is recommended for use in organic analysis. It is included in the Quick-Change Torch Module for HF Analysis (10136-318), the Organics Sample Introduction Kit (10136-316), and the Quick-Change torch module for oil analysis for Optima 4300/5300/7300 V.