Quartz Injector, Int. Ø 2.0 mm Optima 2/4/5/7x00 DV, PerkinElmer
Supplier: PerkinElmer
N0775014
97012-852EA 499.23 USD
97012-852
Quartz Injector, Int. Ø 2.0 mm Optima 2/4/5/7x00 DV, PerkinElmer
Microinjectors Microinjectors, Programmable
This quartz injector is for use with Optima® 2x00/4x00/5x00/7x00 DV ICP-OES series instruments. A variety of sizes are available as an alternative to the standard injector.
Quartz injectors are designed for organic solvents and non-HF matrices.