Alumina Injector for Optima™ 3x00 XL/3x00 DV/3000 SCX/4300V/5300V/7300V, PerkinElmer
Supplier: PerkinElmer
N0695362
97012-836EA 631.5 USD
97012-836
Alumina Injector for Optima™ 3x00 XL/3x00 DV/3000 SCX/4300V/5300V/7300V, PerkinElmer
Chromatography Supplies Chromatography Injectors
This Alumina Injector is used with Optima™ 3x00 XL/3x00 DV/3000 SCX/4300V/5300V/7300V ICP-OES series instruments.
  • Alumina injectors are corrosion-resistant to all mineral acids including HF and aqua regia
  • 2.0 mm Int.Ø
This injector is included in the Quick-Change Torch Module for HF Analysis for Optima™ 4300V/5300V/7300V.