HGA Graphite Tubes with Integrated Platform, PerkinElmer
Supplier: PERKINELMER U.S. LLC
B3001262 B3001264
97012-828PK 1195.85 USD
97012-828 97012-830
HGA Graphite Tubes with Integrated Platform, PerkinElmer
Tubes Graphite Tubes
Graphite tubes with integrated platforms are machined from one piece of graphite, and are totally pyrocoated, providing exceptional tube-to-tube reproducibility.
  • Spherically shaped platform accommodates up to 50 μl of sample
  • Offer an extra measure of convenience versus manually inserting a separate platform
  • Provide longer life with many aggressive sample matrices, such as concentrated acids
Tubes provide STPF conditions for volatile and nonvolatile refractory elements due to the significantly higher heating rates of tube and platform, which can beMore Product Information