MIR Performance Validation Kit, PerkinElmer
Supplier: PerkinElmer
L1202057 L1250404
97012-806EA 61.69 USD
97012-806 10133-738
MIR Performance Validation Kit, PerkinElmer
Chromatography Supplies Chromatography Spare Parts
Instrument performance validation kits are available for both Mid-IR and Near-IR FT-IR instruments
Upgrade kits are available to replace the polystyrene TRM once the validity period has expired.
Delivery information: Kits include a polystyrene Traceable Reference Material (TRM) for abscissa calibration valid for three years, NG11 Schott Glass Reference Material (RM).