Graphite Furnace Autosampler Sample Cups, PerkinElmer
Supplier: PerkinElmer
B0087056 B0510397 B3001566 B3001567 B0087600 B0129303
97012-160PK 231.25 USD
97012-160 97012-776 97012-778 10136-174 10138-980 10138-998
Graphite Furnace Autosampler Sample Cups, PerkinElmer
Sample Containers Sample Cups for Analyzers
These disposable sample cups feature heavy-duty construction to prevent cracking or tipping
  • Polystyrene cups are clear and recommended for use with most aqueous solutions
  • Polyethylene cups are translucent and low-density and recommended for use with most aqueous solutions
  • PTFE cups are recommended for use with most concentrated acids including HF, and for analyses where the lowest detection levels are required
  • Polypropylene cups are translucent and preferred for most solvents
Their conical design allows for use of nearly the entire sample.