PTFE Tubing Assembly, 1 Meter Length, PerkinElmer
Supplier: PerkinElmer
B0191060
97012-692EA 103.1 USD
97012-692
PTFE Tubing Assembly, 1 Meter Length, PerkinElmer
Tubing Transfer Tubing
PTFE tubing assembly includes 1 m length of tubing with 1.0 mm ID, and two blue screw fittings.
This tubing assembly is included in the FIAS standard sampling stainless steel probe assembly, 1.0 mm ID (97013-190) and the FIAS standard sampling probe assembly, 1.0 mm ID (97013-184). A 300 mm PTFE tubing assembly (97012-696) and 700 mm PTFE tubing assembly (97012-698) are available separately.