Flame Capillary Tubing Assembly for S10 and AS-9X Autosampler Probes, PerkinElmer
Supplier: PerkinElmer
B3000157
97012-688EA 143.45 USD
97012-688
Flame Capillary Tubing Assembly for S10 and AS-9X Autosampler Probes, PerkinElmer
Tubing Transfer Tubing
Replacement capillary for the PerkinElmer flame autosampler probe
  • One red male screw fitting (1/4-28 UNF threads)
The capillary is compatible with sample probes for the PerkinElmer S10 and legacy AS-9X autosamplers (AS-90, AS-90A, AS-90 Plus, AS-91, and AS-93 Plus) .