Solvent Flex Peristaltic Pump Tubing Kit, PerkinElmer
Supplier: PerkinElmer
N0680376
97012-678PK 268.75 USD
97012-678
Solvent Flex Peristaltic Pump Tubing Kit, PerkinElmer
Tubing Transfer Tubing
This Solvent Flex Tubing Kit is designed for use with standard pumps on ICP-OES and ICP-MS instruments.
Includes 1.14 mm Int.Ø. Red/Red solvent flex tubing and PTFE tubing for connection to the nebulizer capillary tubing for a 2 ml/min flow rate.
Delivery information: Kit includes: Solvent flex two-stop pump tubing - Red/Red, 1.14 mm I.Ø. (97012-042) (12) and PTFE adapter tubing, (97012-040) (6).