PVC Peristaltic Pump Tubing Kit, PerkinElmer
Supplier: PerkinElmer
N0680375
97012-676PK 126.28 USD
97012-676
PVC Peristaltic Pump Tubing Kit, PerkinElmer
Tubing Transfer Tubing
This PVC Tubing Kit is designed for use with standard pumps on ICP-OES and ICP-MS instruments. Includes Int. Ø: 1.14 mm Red/Red Standard PVC Tubing and PTFE tubing for connection to the nebulizer capillary for a 2 ml/min flow rate.
Delivery information: Kit includes: PVC Two-Stop Pump Tubing - Red/Red, 1.14 mm I.Ø. (10136-164) (12), PTFE Adapter Tubing (97012-040) (6), This tubing kit is included in the Hydride Accessory Kit for Avio 200/500.
Order Now
SPECIFICATIONS
Environmentally Preferable