Copper Reagent for Nitrogen Analysis, PerkinElmer
Supplier: PerkinElmer
N2411527
97012-614EA 990.06 USD
97012-614
Copper Reagent for Nitrogen Analysis, PerkinElmer
Analyzers Elemental Analysers
Copper reagent used in PerkinElmer Elemental Analysis (EA) analyzers for the analysis of elemental Nitrogen.
The best way to assure consistent, accurate results from the PerkinElmer Organic Elemental Analyzer is to use only the highest grade of reagents.