Thermocouple Assembly, PerkinElmer
Supplier: PerkinElmer
03190253
97012-596EA 1501.22 USD
97012-596
Thermocouple Assembly, PerkinElmer
Temperature Probes
Thermocouples are industrial assemblies supplied with or without a temperature transmitter.
  • Customizable over a wide range of functions
  • Provided with or without a temperature transmitter
  • Durable
  • Accurate transmission of temperature readings
An extensive range of elements, connection heads, insertion lengths, neck lengths, and process connections can be individually selected for the appropriate application
Thermocouples can be inserted directly into a process or combined with a variety of thermowell designs.
Replacement sensors can also be configured for certain models.