PTFE Tubing, PerkinElmer
Supplier: PerkinElmer
09908689
97012-582EA 303.25 USD
97012-582
PTFE Tubing, PerkinElmer
Tubing Transfer Tubing
Flexible PTFE tubing for use in high temperature furnace exhaust lines.