Capillary Sampling Probe for PerkinElmer AA Graphite Furnace Autosamplers, PerkinElmer
Supplier: PERKINELMER U.S. LLC
B0129258
97012-448PK 757.04 USD
97012-448
Capillary Sampling Probe for PerkinElmer AA Graphite Furnace Autosamplers, PerkinElmer
Electrodes Other Electrodes
Replacement sampling capillary for Atomic Absorption (AA) instruments equipped with a Graphite Furnace. The PTFE probe is capable of pipetting a variety of corrosive matrices and organic solutions.
  • Works well with solvent and acid-resistant sampling components
  • PTFE pipette tips and capillaries reduce contamination
  • Easy to replace
Compatible with the following, Graphite Furnace Autosamplers: AS-60, AS-70, AS-71, AS-72, AS-800, and AS-900 series.