Aluminium Sample Pans, PerkinElmer
Supplier: PerkinElmer
B0143021 02190041
97012-266PK 833.91 USD
97012-266 97013-054
Aluminium Sample Pans, PerkinElmer
Trays/Dishes/Pots Laboratory Trays/Dishes
Each sample pan and cover has an operating range of -170° to 600 °C, and an ambient max pressure.
  • Can be used with nonvolatile solid samples
  • Crimped design
  • Not hermetically sealed
These sample pans can be used with nonvolatile solid samples, such as plastics and polymers, and are routinely used for applications such as polymer melting, crystallization, glass transitions of polymers, thermoplastics, and thermosets. Pans are crimped, but not hermetically sealed. Not for use with Diamond DSC, PYRIS 1 DSC, or DSC 7 Autosamplers.