Waste and Wash Vials for Autosampler, PerkinElmer
Supplier: PerkinElmer
09923031
97012-220EA 7.61 USD
97012-220
Waste and Wash Vials for Autosampler, PerkinElmer
Vials
These vials are manufactured from Type I Borosilicate Glass, which meets all USP, JP, and EP Pharmacopeia requirements.
  • Excellent at high temperatures
  • Chemical resistant to acidic, neutral and alkali solutions
All vials are packed in a clean environment to ensure you receive contaminant free product every time.