Alumel Calibration Reference Material, PerkinElmer
Supplier: PerkinElmer
09988015
97012-174EA 209.44 USD
97012-174
Alumel Calibration Reference Material, PerkinElmer
Standards Alloys and Metal Ore Standards
Magnetic transition temperature of 154.2 °C, 3 x 2" pieces of alumel wire.