PTFE Tubing, Standard, PerkinElmer
Supplier: PerkinElmer
B0029792
97012-052EA 21.35 USD
97012-052
PTFE Tubing, Standard, PerkinElmer
Tubing Transfer Tubing
This PTFE Tubing can be used to prepare tubing assemblies for AA instruments.
This tubing is 1 meter in length and has a 1.0 mm inner diameter.