Standard PVC 2-Stop Pump Tubing, PerkinElmer
Supplier: PerkinElmer
09908587 N0695476 N8122012
97012-050PK 70.59 USD
97012-050 97012-652 97012-668
Standard PVC 2-Stop Pump Tubing, PerkinElmer
Tubing Transfer Tubing
PVC Flow-measured tubing provides excellent pumping life, good handling characteristics, and low permeability.
  • Excellent pumping life
  • Low permeability
  • Color-coded stops
  • Also available with flared ends
Tubing has an overall length of 455 mm/18" with two color-coded stops separated by 152 mm/6". All PVC pump tubing is also available with flared ends.