Palladium 1% (Pd) matrix modifier solution, for graphite furnace AAS

Supplier: PerkinElmer

B0190635
97012-036EA 760.71 USD
97012-036
Palladium 1% (Pd) matrix modifier solution, for graphite furnace AAS
Palladium

This 1% Pd as nitrate matrix modifier is used for Atomic Absorption (AA) THGA graphite furnaces.

Formula: Pd
MW: 106.420 g/mol
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR