Lead Standard Solution, for Atomic Grade Spectroscopy, PerkinElmer
Supplier: PerkinElmer
Pure Grade Single element Standards for AA and ICP-OES.
- Matrix of 2% HNO3
- Concentration of 1000 µg/mL
Certifications: ISO 17034 and ISO 17025, certified by A2LA
Delivery information: Supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...