Multi-Element Quality Control Standard, 21 Elements, PerkinElmer
Supplier: PerkinElmer
N9300281
97011-998EA 634.77 USD
97011-998
Multi-Element Quality Control Standard, 21 Elements, PerkinElmer
Standards Cationic Standards Multi-Element Standards
Use for standard metal analysis.
  • Matrix: 5% HNO3
  • Liquid format
21 elements (100 mg/L): As, Be, Ca, Cd, Co, Cr, Cu, Fe, Li, Mg, Mn, Mo, Ni, Pb, Sb, Se, Sr, Ti, Tl, V and Zn.