Instrument Calibration Standard 4 for CLP, PerkinElmer
Supplier: PERKINELMER U.S. LLC
N9300221
97011-988EA 408.27 USD
97011-988
Instrument Calibration Standard 4 for CLP, PerkinElmer
Standards Cationic Standards Multi-Element Standards
Solution needed for daily calibration of ICP and HGA for CLP analyses.
Recommended dilution: a 100-fold dilution with high-quality 5% HNO₃. Remember to prepare your calibration standards fresh for each analysis and discard after use.