Setup/Stab/Masscal Solution for ELAN 9000/6100, PerkinElmer
Supplier: PerkinElmer
N8125030
97011-972EA 319.84 USD
97011-972
Setup/Stab/Masscal Solution for ELAN 9000/6100, PerkinElmer
Standards Cationic Standards Multi-Element Standards
Inorganic solution is for use with the ELAN 9000/6100 in atomic spectroscopy.