Lab Digital Refractometer, Brix 45 - 95%, Sper Scientific
Supplier: Sper Scientific
300033
97011-856EA 1004.94 USD
97011-856
Lab Digital Refractometer, Brix 45 - 95%, Sper Scientific
Refractometers Benchtop Refractometers
Measure concentrations in brix, salinity, refractive index, and clinical parameters with the full scale accuracy of 0.1% using CCD scanner (linear scanned array imaging) technology
These simple to use models calibrate to distilled water and require a sample of only 0.4mL. Instantly displays temperature in addition to multiple scales with the unit of measure indicated on the large LCD. The sophisticated microprocessor provides automatic calibration, automatic temperature compensation, automatic shutoff (after 3 minutes), and visual min./max. alarms. Low power consumption enables 5,000 readings, before the LCD indicates that the battery should be replaced. The water-resistant design protects the electronics from wet environments and allows for easy clean ups.
Ordering information: Supplied ready to use with a 9V battery, distilled water, transfer-pipet, instructions, and a protective case. The refractometer also includes a 1-year warranty.