Dissolved Oxygen Pen, Sper Scientific
Supplier: Sper Scientific
850045
97011-782EA 950.73 USD
97011-782
Dissolved Oxygen Pen, Sper Scientific
Electrochemistry Meters Oxygen Meters
Easy to carry, simple to calibrate and use, and sufficient for many surface dissolved oxygen measurements.
Measures dissolved oxygen to determine water quality in environmental testing, labs, industrial and municipal waste water, aquariums and fish hatcheries. Designed for simple one-hand operation with automatic temperature compensation and calibration to air. The polarographic probe has a built in temperature sensor and easy-to-replace pop-off probe heads. Simultaneously displays dissolved oxygen and temperature readings. Also holds readings and recalls minimum and maximum. IP67 waterproof.
Ordering information: Comes ready to use in a soft carrying case with belt loop, two spare probe heads, electrolyte and 4 AAA batteries. Five year warranty.