VWR® 5000IR Incubating/Refrigerating Orbital Shaker, 230 V (Export Only)
Supplier: VWR International
80863571
97009-896EA 18839.43 USD
97009-896
VWR® 5000IR Incubating/Refrigerating Orbital Shaker, 230 V (Export Only)
Shakers and Mixers Shaking Incubators
Shaker offers a diverse combination of advanced operating and safety features for reliable, high-performance shaking and incubating applications in cell cultures, solubility studies, bacterial suspensions, and extraction procedures. 
  • 230 V model
  • Exclusive opti-flow forced ventilation system
  • Exclusive Accu-Drive™ shaking system
  • Best-in-class, five-year warranty
  • Available with a NIST Traceable, ISO/IEC 17025 Accredited Calibration Certificate from Troemner
Variable speed microprocessor control provides consistent and uniform mixing action with the ability to restart and display last set-point if power is interrupted
A variety of safety features are standard on the Model 5000I/R shaker. A built-in load sensor detects unbalanced conditions and reduces rpm or stops the unit to protect samples. Audible and visual signals will activate if the system detects a tray obstruction or overload. To avoid sample loss, a ramping feature slowly increases speed to setpoint. In timed mode, an alarm will sound when the timer reaches the zero point. A polycarbonate lid permits viewing of samples without disturbing internal temperature and, when opened, an automatic shutoff circuit will stop the platform operation. Each unit's spill-resistant design protects internal components from accidental spills and leaks. Tray and rubber mat are removable to allow for easy chamber cleaning. A caution symbol will illuminate when air temperature in the chamber reaches 40 °C (104 °F). This symbol remains lit until the chamber temperature cools. 
The touchpad control features easy-to-read, independent LEDs for temperature, time, and speed. An RS-232 interface provides two-way connection for data logging and unit control. 
Precise temperature control is maintained by a PID controller, and the unit is driven by a triple eccentric drive. Permanently lubricated bearings and a maintenance-free brushless DC motor ensure reliable service and continuous duty operation. Shaker is equipped with the Accu-Drive™ shaking system, which delivers exceptional speed control, accuracy, and durability. The system continuously monitors shaking speed and maintains setpoint even under changing loads. The displayed speed is accurate to 1% of setting. The Opti-Flow ventilation system utilizes a powerful fan and air deflectors to deliver optimal temperature uniformity. Built-in temperature calibration mode and speed calibration mode allow users to calibrate temperature to an external device, or to automatically recalibrate the speed display.
Certifications: RoHS certified. CE marked, TUV listed. Calibrated shakers include a Troemner ISO/IEC accredited calibration certificate traceable to NIST. The ISO/IEC 17025 accredited calibration certificate includes multiple data points within the speed range and time function with the associated uncertainties. Troemner is accredited by NVLAP under laboratory code 105013-0 to perform ISO/IEC 17025 calibrations.
Ordering information: Shaker includes a non-skid rubber mat that fits the 45.7×45.7 cm (18×18") tray. Tray is suitable for holding Petri dishes and cell culture flasks. Units also includes a 92" (234 cm) Euro type plug. Optional stainless steel platform can be used for mounting Erlenmeyer flask clamps and test tube racks.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (97009-894) is also available, please see separate product page.