Lead-Free, Non-Aerated Eyesaver Faucet, Model SEF-1800NA, Speakman®
Supplier: Speakman
SEF-1800-CA-NA
97008-324EA 1610.95 USD
97008-324
Lead-Free, Non-Aerated Eyesaver Faucet, Model SEF-1800NA, Speakman®
First Aid Supplies Eyewash Stations and Eyewash Bottles
Speakman® Eyesaver® combination eyewashes and lavatory faucets are a revolutionary and exclusive product with a unique design.
  • The faucet always remains functional
It combines a fully functional gooseneck faucet with an independently operated eyewash. This patented design incorporates two separate water channels in the faucet body, one for the eyewash and the other for the faucet. A simple pull activates the eyewash and delivers a safe water temperature to the eyes each and every time. The SEF-1800NA Model is a two-handle, widespread, chrome-plated, lead-free brass gooseneck faucet with integral non-aerated eyewash spray outlets.
Certifications: Meets ANSI Z358.1, NSF 61, CA AB1953, and VT Act 193.