OIML Class F2 Precision Weight Sets, Mettler Toledo
Supplier: Mettler Toledo
30406439
97007-746EA 1661.63 USD
97007-746 97018-752 97018-754 97018-756 97018-758 97018-760 97018-762 97018-750
OIML Class F2 Precision Weight Sets, Mettler Toledo
Weights Calibration Weights
Basic Line weights are protected in robust and easy to clean plastic boxes
  • Made of stainless steel
  • Adjustable cavity (AC) design
  • Economical
FDA approved materials, including foam inserts, make them a perfect solution for regulated industries.
Delivery information: All weight sets delivered with tweezers.