OIML Class 4 Individual Precision Weights, ASTM, Mettler Toledo
Supplier: Mettler Toledo
11123584 11123585 11123586 11123587 11123588 11123589 11123590 11123591 11123592 11123593 11123596 11123598 11123601 11123603 11123604 11123608 11123609 11123611 11123613 11123614 11123615 11123616 11123618 11123619 11123620 11123622 11123623 11123624 11123625 11123626 11123594 11123597 11123599 11123602 11123610 11123617 11123627 11123595 11123600 11123607 11123612 11123621 11123628 11123629 11123605 11123606
97007-486EA 55 USD
97007-486 97007-488 97007-490 97007-492 97007-494 97007-496 97007-498 97007-500 97007-502 97007-504 97007-510 97007-514 97007-520 97007-524 97007-526 97007-530 97007-532 97007-536 97007-540 97007-542 97007-544 97007-546 97007-550 97007-552 97007-554 97007-558 97007-560 97007-562 97007-564 97007-566 97007-506 97007-512 97007-516 97007-522 97007-534 97007-548 97007-568 97007-508 97007-518 97007-528 97007-538 97007-556 75852-890 75852-892 75852-894 75852-896
OIML Class 4 Individual Precision Weights, ASTM, Mettler Toledo
Weights Calibration Weights
ASTM weights are designed to resist dust and moisture intrusion, offering long-term stability.
  • 316 stainless steel
  • Low magnetic permeability
  • High corrosion resistance
  • High gloss polish
All precision weights are protected in robust and easy to clean plastic boxes. FDA approved materials, including foam inserts, make them a perfect solution for regulated industries.