CarePac® Test Weight Sets, METTLER TOLEDO®
Supplier: Mettler Toledo
11123100 11123102 11123107 11123147 11123148 11123151 11123153 11123154 11123157 11123158 30550614 30550649 11123009 11123044 11123046 11123047 11123048 11123054 11123105 11123104 11123004 11123101 11123103 11123145 11123152 11123106 11123108 11123146 11123159 30550613 11123045 11123049 11123050 11123051 11123052 11123053 11123056 11123057 11123058 11123059 11123109 11123110 11123150 30550616 11123008 11123055 11123011 11123144 11123149 11123155 11123156 11123010 11123111
97007-250EA 730 USD
97007-250 97007-254 97007-258 97007-272 97007-274 97007-280 97007-284 97007-286 97007-292 97007-294 97007-300 97007-306 97007-310 97007-314 97007-318 97007-320 97007-322 97007-334 97019-006 97019-008 97019-014 97007-252 97007-256 97007-268 97007-282 97019-004 97007-260 97007-270 97007-296 97007-298 97007-316 97007-324 97007-326 97007-328 97007-330 97007-332 97007-338 97007-340 97007-342 97007-344 97007-262 97007-264 97007-278 97007-304 97007-308 97007-336 97027-718 97007-266 97007-276 97007-288 97007-290 97007-312 97027-716
CarePac® Test Weight Sets, METTLER TOLEDO®
Weights Calibration Weights
These sets enable routine testing of balances by using two weights at maximum and minimum loads
  • Routine testing of balances
  • Multiple configurations
Weights are available in both ASTM and OIML configurations and are specified to validate process tolerances up to 0.03%. Sets include ergonomic tweezers and cleaning cloths. Customized third weights are available for customer-specific testing, such as minimum weight determination. Specially-designed cases house both types of weights and all equipment. Additional space is provided to accommodate a third weight.