Catapult™ 1000 Compact Bench Scales, Ohaus®
Supplier: Avantor
97006-726EA 356.98 USD
97006-726 97006-728 97006-730
Catapult™ 1000 Compact Bench Scales, Ohaus®
Balances and Scales
Catapult™ 1000 bench scales offer reliable and precise performance in an economic, compact design.
Three capacities, ranging from 9kg (20lbs.) to 75kg (165lbs.), are available with both weighing and dynamic weighing modes
This bench scale series is ideally suited for home use, general shipping applications, veterinary weighing, and other dry weighing applications.
Overload protection is designed into the weigh cells for use in tough environments where capacity is regularly exceeded. Built-in carrying handles allow for easy transport. Additional standard features include an AC power pack, ABS plastic housing, two raised tactile keys, rubber nonslip feet, and digital external calibration. Scales also provide the option of battery powered operation with a power-saving auto shutoff mode.