Polytron® 12 mm Generator, PT-DA 12/2EC-E157, Kinematica

Supplier: Kinematica
11030060
97006-686EA 3100.31 USD
97006-686
Polytron® 12 mm Generator, PT-DA 12/2EC-E157, Kinematica
Homogenizers Dispersing Tools
Fits Polyron models, PT 1200 E, PT 1300 D, PT 2500 E as well as discontinued model PT 1600 E.

  • Saw tooth stator design for added size reduction effects
  • Easy clean design for quick disasembly
  • 316L stainless

Precison machined form the finest 316 SS by our skilled Swiss craftsman for years of trouble free service.

Certifications: CE marked.

Accessories information: Generators and stand with vessel holder sold separately.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR